NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Proveedor: Air-Tite
NORM-JECT®
8300020454 8300020495 8300020453 8300020456 8300020460 8300027526 8300027527 8300027529 8300020496 8300020494 8300020452 8300020458 8300020493
89174-504CS 21001.37 MXN
89174-504 89213-176 89174-506 89213-182 89174-496 89174-498 89174-500 89213-178 89174-502 89174-492 89174-491 89174-494 89213-180
NORM-JECT® Luer-Lock and Luer-Slip Bulk Syringes, Two-Part, Nonsterile, Air-Tite
Jeringas Jeringas de uso múltiple
Norm-Ject® Syringes are latex free, contain no rubber, no silicone oil, styrene or DEHP and are DNA free.
  • All-plastic construction
  • Low background extractables
  • Wide solvent compatability
  • Positive safety stop to prevent accidental spills
The 5 ml syringes are graduated to 6 ml, 10 ml syringes are graduated to 12 ml, 20 ml syringes are graduated to 24 ml, and 50 ml syringes are graduated to 60 ml.
More biologically inert than standard 3-part syringes. Because of their composition they are indicated for chromatography and many laboratory procedures.
Certificaciones: Meets ISO standards, HPLC certified.
Envase: The syringes are packed bulk, non-sterile in re-closeable plastic bags.