VWR® Wash Bottles, Right-to-Know, Safety-Vented, with GHS Labeling
Proveedor: VWR
VWR®
12416-6001 12416-6007 12432-6015 12416-6016 12432-6001 12432-6004 12432-6050 12416-6002 12416-6021 12416-6020 12432-6008 12416-6003 12432-6019 12416-6013 12416-6010 12416-6019 12416-6017 12416-6008 12416-6004 12416-6012 12416-6015 12416-6050 12432-6011 12416-6011
10111-946CS 6378.93 MXN
10111-946 10111-968 10111-958 10111-948 10111-816 10111-818 10111-982 10111-960 10111-972 10111-950 10111-962 10111-940 10111-952 10111-974 10111-942 10111-964 10111-976 10111-954 10111-944 10111-966 10111-978 10111-956 10111-980 10111-970
VWR® Wash Bottles, Right-to-Know, Safety-Vented, with GHS Labeling
Frascos Frascos de lavado
Safety-vented wash bottles provide compliance with the Globally Harmonized System of Classification and Labeling of Chemicals (GHS) and OSHA’s HazCom 2012 requirements - as pertains to 'Workplace Labels'.
  • Pre-printed with chemical name and formula
  • Unique valve design improves use and safety
  • Translucent LDPE construction with color-coded polypropylene closures
They are pre-printed with the chemical name and formula, GHS pictogram and signal word*, National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment.
A unique valve prevents pressure build-up, eliminating potentially dangerous chemical drips, and allows dispensing in an upright or inverted position. The 53 mm (2") wide mouth makes refilling easy. Bottles are translucent LDPE (Sodium Hypochlorite is white LDPE for light-protection between 230 and 450 nm and Toluene is red LDPE) with color-coded polypropylene closures to aid in quick and easy identification.
Note: *Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word. Methyl Ethyl Ketone and Toluene assemblies do not include vented valve.
Información para pedidos: Assortment orders include four bottles per bag, with one each of, Acetone, Isopropanol, Bleach, and Ethanol.