Face Shields and Visors, 3M™
Proveedor: 3M
7000127234 7000127231 7000052705 7000127236 7000002339 7100000260 7000127232 7000127168 7000052706 7000002337 7000127236 7000127169 7000127231 7000127232 7000052753 7000002339 7000002340 7100000260 7000052706 7000127231 7000002340 7000127241 7000002337 7000127169 7000052705
89006-138CS 27595.26 MXN
89006-122 55666-308 CA56615-616 CA56615-736 80077-247 CA56615-618 55666-310 CA56615-732 56615-725 CA56615-614 89006-138 CA89006-122 CA55666-308 CA47747-276 89107-468 CA56615-725 56615-618 89107-446 CA80077-247 CA55666-310 56615-732 56615-614 CA89006-138 56615-736 56615-616
Face Shields and Visors, 3M™
Protectores faciales
Faceshields provide ventilation and face protection for high heat environments.
  • Faceshieldd mount quickly to the appropriate 3M™ headgear without tools, snaps, or cams
  • Replaceable eyeshields offer increased visibility
Certificaciones: Items meet ANSI Z87.1-2003 standards.