Legato 270 Dual Syringe Push-Pull Programmable Syringe Pump, KD Scientific
788272
10117-892EA 157272.8 MXN
10117-892
Legato 270 Dual Syringe Push-Pull Programmable Syringe Pump, KD Scientific
Push/Pull syringe pump
  • Infuse only dual syringe pump
  • 6400 microsteps per one revolution of lead screws
  • Real-time clock, touch pad "lock" feature, LED light on front panel
  • Unparalleled ease of use
  • Full metal chasis
  • Built-in syringe table
  • Advanced microstepping techniques
  • Built-in RS-485 interface, USB port, I/O and TTL interface
  • Continuous mode of operation, spill dam
Accomodates 4 syringes size 0.5ul to 140ml, simultaneously operates two for infusion and two for withdrawal. High resolutuion touch screen display. User definable flow rates. Programmable with 40 multi-step programs using constant rate, ramp, pulsed & stepped profiles.
Certificaciones: UL, CSA, CUL, CE (Global), RoHS, WEEE.
Precaución: KD Scientific syringe pumps are for laboratory use only. Not for clinical use.