SP Bel-Art H-B® DURAC® Salt Brine Plastic Plain Form Hydrometers, Traceable to NIST, Bel-Art Products, a part of SP
Proveedor: Bel-Art Products, a part of SP
B61800-1000 B61800-0600
89217-457CS 40010.59 MXN
89217-457 89217-449
SP Bel-Art H-B® DURAC® Salt Brine Plastic Plain Form Hydrometers, Traceable to NIST, Bel-Art Products, a part of SP
Hidrómetros
Made from optical quality polycarbonate plastic material, these plain form hydrometers are durable and shatterproof, and perfect for field work where breakage could be a concern.
  • Polycarbonate plastic construction
  • Durable and shatterproof
  • Easy-to-read black scale on yellow back
  • Individually serialized
They are used for measuring sodium chloride saturation of salt brines in various industries including food, fishing, and transportation
Note: Polycarbonate hydrometers are not recommended for use with petroleum-based liquids or other corrosives that would potentially affect the plastic material. They should be stored in a clean, dry, safe place and should not be subjected to extreme temperatures.
At 300 mm (12”) long, they contain lead free ballast and are available in models to measure various levels of salt content from 0% to 100%. Easy-to-read black scale on yellow back helps avoid eye fatigue. Each is individually serialized and has a temperature standardization of 60°F (15°C).
Certificaciones: DURAC® Polycarbonate Hydrometers are made and hand inspected by our U.S. quality team in our exclusive triple accredited/registered ISO/IEC 17025:2005 and A2LA Accredited Calibration Laboratory; internationally recognized and accepted by the signatory economies of the ILAC/MRA and located in our ISO 14001:2004 Environmental Management System registered facility. They are packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Made in USA.