Datalogging Thermometer, 12-Channel, Extech
Proveedor: Extech
TM500
10068-326EA 38949.69 MXN
10068-326
Datalogging Thermometer, 12-Channel, Extech
Registradores de datos
Extech’s 12-Channel datalogging thermometer allows for a wide-range of applications with 6 Thermocouple types (J, K, E, T, R,S)
Offset adjustment used for zero function or to make relative measurements. Unit stores 99 readings manually and 20M readings via 2GB SD card.
It displays CH1 to CH8 or CH9 to CH12 on screen, and the data can be recorded from all 12 channels simultaneously onto an SD card in Excel format for further analysis.
Certificaciones: CE