VWR® Gram Stain Sets and Reagents
Proveedor: VWR
VWR®
6255 6254 6501 6251 6928 6552 6253 6505 6930-G 6932-G 6504 6600 6506 6256 6933-G 6931-G 6929-G 6400
10143-178CS 5425.06 MXN
10143-170 10143-182 10143-180 10143-174 10143-262 10143-252 10143-172 10143-260 10143-184 10143-178 10143-168 10143-256 10143-176 10143-264 10143-254 10143-258 10143-604 10143-602
VWR® Gram Stain Sets and Reagents
Tintes Tintes para microbiología
Gram Stain Sets include all the necessary solutions to fully differentiate Gram positive and Gram negative specimens.
  • Sets include all necessary solutions
  • Enables differentiation
All Gram stain and replacement sets are packaged in a styrofoam tray and come with spout caps for ease of pouring.