VWR® PCR Storage Rack
Proveedor: Avantor
2721-229-000
83009-690PK 1722.93 MXN
83009-690
VWR® PCR Storage Rack
Gradillas Tube Racks
The polypropylene 96-well PCR storage rack is perfect for storing samples before cycling or working with completed procedures.
  • Racks hold tubes, strips, or plates that have a separate snap-on lid
  • Autoclavable and freezable
Designed for preparing and storing thin-walled PCR tubes and strip tubes before or after PCR sample processing applications.
Información para pedidos: Available in assortment pack of five colors: red, orange, yellow, green and blue.