VWR® Ice Buckets with Lids
Proveedor: Avantor
VWR®
525071B 525071BK 525071R 525131G 525131R 525131B 525131BK 525071G
10146-188EA 2216.26 MXN
10146-190 10146-188 10146-184 10146-186 10146-192 10146-290 10146-194 10146-292
VWR® Ice Buckets with Lids
Contenedores de hielo
VWR® Ice Buckets with Lids are manufactured from polyurethane foam to ensure good insulating properties.
  • Polyurethane foam construction
  • pouring lip for easy draining
  • Lids can be stacked beneath buckets
The Ice Buckets are suitable for use with Dry-Ice with Isopropyl Alcohol, Water-Ice with salt slurry, or liquid nitrogen. The operating temperature range is -76 to 70°C (-104 to 158°F). All buckets feature a pouring lip for easy draining. The lids, which are unbreakable in normal use, help to reduce evaporation levels. Lids can also be stacked below buckets for convenient, space-saving storage.