Biotube™ Cluster Tube System, Simport
Proveedor: Simport
T100-60R T100-60R T100-3B T100-2B T100-60B T100-50B T100-1Y T100-2Y T100-1R T100-20LST T100-50B T100-26 T100-1B T100-4G T100-2R T100-4Y T100-60B T100-4R T100-3R T100-26 T100-28 T100-1G T100-60G T100-1Y T100-3Y T100-4B T100-3R T100-35 T100-4B T100-40 T100-2G T100-3Y T100-1G T100-25 T100-2Y T100-20 T100-30 T100-4R T100-1R T100-25 T100-4Y T100-50G T100-3G T100-2R T100-2B T100-50G T100-28 T100-3G T100-4G T100-3B T100-35 T100-1B T100-20
60872-720CS 2999.4 MXN
10199-558 CA60870-021 10199-630 CA60872-504 60872-698 CA60872-509 CA60872-506 CA60872-508 CA60872-507 60870-022 60870-026 10199-640 60872-694 60872-696 60872-690 60872-692 60872-720 60872-686 60872-722 60872-688 CA60870-026 CA60870-028 CA60872-494 60872-684 CA60872-490 CA60872-690 CA60872-524 60872-712 CA60872-688 CA60872-526 60872-714 CA60872-520 CA60872-684 CA60872-489 60872-710 CA60872-686 CA60872-521 60872-716 60872-718 10199-548 10199-546 10199-544 60872-702 CA60872-513 CA60872-512 CA60872-515 60872-704 CA60872-514 60872-700 CA60872-511 CA60872-510 60872-706 60872-708
Biotube™ Cluster Tube System, Simport
Tubos Microtubos
The Biotube™ Cluster Tube System is a polypropylene tube and rack system with tubes held in a standard 8×12 configuration.
  • Alphanumeric coding and removeable 96-place ID card for accurate sample location
  • Transparent cover keyed to base to prevent misalignment
  • Ideal for storing, freezing and transporting reagents and specimens
  • Racks and tubes can be used over a wide temperature range, −80 to 121 °C
Tubes and racks can be autoclaved up to 100 times and will resist most research chemicals. The tube supporting grid plate can be removed and used as a self-standing support. It can also be placed in a refrigerator or freezer for improved air circulation around the tubes, or in water baths to allow controlled warming of the tubes contents.