Single Edge Razor Blades, Accutec Blades
Proveedor: AccuTec Blades
62-0178-0000 62-0176-0000 62-0177-0000 62-0179-0000
10040-384CS 28306.74 MXN
10040-386 10040-380 10040-384 10040-382
Single Edge Razor Blades, Accutec Blades
Cuchillas
Single Edge blades provide a solution for virtually any need
Each blade is individually wrapped with a protective shell for added safety. Blades come with aluminum back and have a triple facet edge for critical cuts that enhance productivity and performance.