DURAN® Laboratory Bottles with GL Thread, Ace Glass
Proveedor: Ace Glass
DURAN®
5539-149 5539-235 5539-35 5539-41 5539-215 5539-84 5539-118 5539-66 5539-258 5539-62 5539-90 5539-208 5539-262 5539-81 5539-86 5539-25 5539-79 5539-230 5539-254 5539-156 5539-150 5539-05 5539-15 5539-50 5539-60 5539-88 5539-264 5539-229 5539-64 5539-256 5539-222 5539-22 5539-205 5539-130 5539-52 5539-125 5539-49 5539-56 5539-70 5539-122 5539-154 5539-260 5539-80 5539-250 5539-115 5539-85 5539-18 5539-91 5539-249 5539-266 5539-241 5539-03 5539-268 5539-68 5539-10 5539-252 5539-83 5539-58 5539-158 5539-08 5539-218 5539-160 5539-82 5539-244 5539-54 5539-44 5539-152 5539-30 5539-105 5539-210 5539-89
89051-835CS 8601.51 MXN
89051-835 89051-832 89051-798 89051-830 89051-796 89051-795 10098-788 89051-792 89199-038 10098-786 89051-790 89214-894 89199-030 10098-780 89214-892 89199-032 89214-898 89199-034 10098-784 89214-896 10098-782 89199-036 89051-828 89199-060 89051-826 89051-824 89051-788 89051-786 89051-784 89199-028 10098-778 89051-782 10098-776 89051-780 10098-770 89199-020 89199-022 89199-024 10098-774 89199-026 10098-772 89051-818 89051-816 89199-050 89051-812 89051-778 89199-016 89199-018 10098-768 89199-052 89199-010 89199-054 89199-056 89199-012 89199-014 89199-058 89051-808 89051-804 89199-040 89051-802 89051-845 89051-800 89051-843 89051-841 89199-006 89199-008 89199-042 10098-790 89199-044 89199-046 89199-048
DURAN® Laboratory Bottles with GL Thread, Ace Glass
Frascos Botellas de vidrio
DURAN® graduated laboratory bottles are chemically resistant and stable.
Note: 10 and 25 ml sizes do not have pouring rings.
Bottles are available with or without red PTFE-lined PBT cap and a removable pouring ring that allows for drip-free operations. Bottles are designed for use in temperatures up to 180 °C (356 °F). Blue polypropylene caps are suitable for use in temperatures up to 140 °C (284 °F). Not suitable for vacuum or pressure use. Bottles can be hot air sterilized or autoclaved. Amber colored bottles offer protection to samples which are sensitive to light. Safety coated bottles offer protection against mechanical damage and prevent leakage of contents should the glass break. The safety coated bottles are suitable for use up to 135 °C (275 °F). Not suitable for vacuum or pressure use. All bottles are fabricated from borosilicate glass.