Whatman™ Grade 598 Qualitative Filter Papers, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™
10312287 10312209 10312287 10312000 10312209
10034-684 CA74330-470 76322-126 CA11012-762 74330-470
Whatman™ Grade 598 Qualitative Filter Papers, Whatman products (Cytiva)
Filtros Papeles de filtros Papel de filtro cualitativo
A thick filter paper with high loading capacity. Combines medium retention with medium-fast to fast filtration speed.
These cellulose filters are used in qualitative analytical techniques to determine and identify materials. Prepleated qualitative filters are also available, which give improved flow rate and increased loading capacity compared to equivalent flat filters.