SP Bel-Art Lab Drawer Compartment Trays, Bel-Art Products, a part of SP
Proveedor: Bel-Art
SCIENCEWARE®
H18655-0000 H18648-0000 H18650-0000 H18654-0000 H18658-0000 H18652-0000
70680-528CS 13175.59 MXN
70680-526 70690-190 70680-528 70680-160 70680-136 70680-158
SP Bel-Art Lab Drawer Compartment Trays, Bel-Art Products, a part of SP
Bandejas/Platos/Ollas Bandejas para organización de cajones
These white polystyrene plastic trays measure 36 x 44.5 x 5.7 cmH (14 x 17½ x 2¼”) and fit standard lab bench drawers.
  • White polystyrene construction
  • Fits standard lab bench drawers
  • Easy and convenient organization
They are a practical and functional solution for organizing the “stuff” of daily lab life including thermometers, medication cups, jars, stir bars, vials, and pipettes.