Eisco® NextGen Tirrill Bunsen Burner
Proveedor: EISCO SCIENTIFIC LLC
NextGen™
CH0995LP CH0995NG
470121-708EA 1033.34 MXN
470121-708 470121-724
Eisco® NextGen Tirrill Bunsen Burner
Calentadores Mecheros de gas
The next generation in laboratory burners. NextGen Bunsen Burners are designed to be the safest on the market.
  • Designed for safety
  • Expanded base prevents easy tipping, even with hoses connected
  • Spaced feet so burner fits comfortably under stands
  • Candle stick style handle allows user to move burner without getting harmed
  • Equipped with gas adjustment under the burner and threaded air flow adjustment
  • Includes brass flame stabilizer
  • Base is coated with a chemical resistant powder coating
This Bunsen burner produces a safe continuous stream of flammable gas used for heating, sterilization and combustion. The Tirrill is a specially designed type of Bunsen burner that features a wing nut on the bottom of the tube for gas adjustment and the tube screws for fine adjustment of the air to fuel ratio. This is a very safe and easy to use burner. Ribbed fuel input tube is 8 mm outside diameter, and versions jetted for liquid propane and natural gas are both available. Liquid propane burners have a BTU range of approx. 2000 to 3200. Maximum temperature is approximately 1550 °C (2822° F). Natural gas burners have a BTU range of approx. 800 to 1200. Maximum Temperature is approximately 1500 °C (2732° F).
Información para pedidos: Neoprene connecting hoses, 60 or 90 cm (items 470121-742 and 470220-806), recommended.