VWR® Mini Hotplate 120 V
Proveedor: VWR
VWR®
80863876 80863878
10153-312EA 5566.58 MXN
10153-312 10153-300
VWR® Mini Hotplate 120 V
Calentadores Placas calefactoras
Rugged, compact hotplates with an ergonomic design are ideal for educational labs and can heat up to 1000 mL of water.
  • Rugged 5x5" aluminum top plate
  • Boils 300 mL of water in 16 minutes
  • Built-in support rod holder
  • Five-Year Warranty on Parts and Labor
Durable 5x5" cast aluminum top plate will not chip or crack, and provides a uniform heating surface
The fixed temperature hotplate features an illuminated rocker switch to activate the preset fixed temperature of 375°C.
The built-in support rod holder with locking knob accepts an optional support rod and clamp kit. Kit includes 45.7 cm (18”) support rod, thermometer extension clamp, three-prong medium swivel clamp, and hook connector. Optional support plate allows hotplate to be mounted to lab frames or ring stands.
Bi-metallic thermostat offers reliable temperature control. Powerful heater reaches maximum temperature in minutes.
Certificaciones: TUV listed. CE marked. RoHS certified.
Información para pedidos: The 120V units include a 183 cm (72"), 3-wire cord and plug. All units include a five-year limited warranty on parts and labor.