Whatman™ 3-Piece Filter Funnels, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™
1950-007 1950-012 1950-027 1950-027 1950-002 1950-017 1950-004 1950-012 1950-007 1950-002 1950-009 1950-017 1950-004
28200-200EA 11393.28 MXN
CA11013-674 CA11013-210 28200-200 CA80085-988 28200-254 80085-988 CA28200-200 CA80085-986 28200-301 CA28200-254 80085-986 10035-336 10035-334
Whatman™ 3-Piece Filter Funnels, Whatman products (Cytiva)
Embudos Embudos de filtración Embudos con filtro
Whatman 3-piece filter funnels from Cytiva are ideal for quick and easy filtration using high-efficiency glass microfiber filters.
  • Chemically resistant - 3-piece construction consists of glass cylinder and base with a polymeric membrane support
  • Support options - Provided with acrylic support and optional polypropylene or PTFE supports available separately for some sizes
  • Secure sealing - Clamps at edges to prevent peripheral loss due to fluid bypass around the edge of the filter
  • Eliminate carry-over - Simplicity of design facilitates easy and thorough cleaning between use
The increased use of high-efficiency glass microfiber filters in modern laboratories has created a demand for simple and effective filter-holding systems. Whatman 3-piece Filter Funnels have been designed to complement the range of Whatman fine particle retention, rapid flow rate glass microfiber filters.
The unit consists of a glass funnel and base, acrylic support platform, and 3 easy-to-apply metal clips. The range of diameters, funnel sizes, reservoir volumes and alternate support platforms provides a flexible system to meet the varied and unique needs of busy laboratories.