TLC Glass Plates, Silica Gel 60, MilliporeSigma
Proveedor: MilliporeSigma
1.00390.0001 1.00390.0001 1.00384.0001 1.00384.0001
52428-716EA 7865.24 MXN
CA1.00390.0001 52428-716 52428-714 CA1.00384.0001
TLC Glass Plates, Silica Gel 60, MilliporeSigma
Placas TLC
These new TLC silica gel 60G plates are recommended for customers in QA/QC labs using older Ph Eur monograph methods, which require TLC plates with gypsum binder.