Whatman™ ReZist Air Venting Filter, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™ ReZist™
10463607 10463607 10463609 10463608 10463609 10463608
10035-168PK 7200 MXN
CA76423-782 10035-170 CA11013-166 76423-782 10035-168 CA11012-966
Whatman™ ReZist Air Venting Filter, Whatman products (Cytiva)
Filtros Filtros en línea
ReZist in-line filters are lightweight air venting units suitable for protective vents, in-line filtration, and isolation applications when working with chemically harsh solvents.
  • Integral hydrophobic PTFE venting membrane is resistant to harsh solvents
  • Can be autoclaved multiple times without loss of integrity
  • Integrity-testable (bubble point or water breakthrough pressure in situ methods)
  • Lightweight: avoids the collapsing of tubing that can be caused by heavy filter devices
  • Appropriate for in-line HPLC sample preparation
This device features a PTFE venting membrane, which is well suited for chemically aggressive solutions, reagents, and organic solvents.