Buffer tablets for Haematology to prepare solutions according to Wiese, MilliporeSigma
Proveedor: MilliporeSigma
1.11373.0100 1.09468.0100 1.11374.0100 1.11374.0100 1.09468.0100
EM1.09468.0100 CA1.11374.0100 EM1.11374.0100 CA1.09468.0100 EM1.11373.0100
Buffer tablets for Haematology to prepare solutions according to Wiese, MilliporeSigma
Tampones Tampones para microscopía
Used to prepare buffer solution acc. to WEISE for staining blood smears.