Brix Sugar Standards for refractometers, Certipur, MilliporeSigma
Proveedor: MilliporeSigma
5.00410.0001 5.00450.0001 5.00430.0001 5.00420.0001 5.00400.0001 5.00420.0001 5.00430.0001 5.00405.0001 5.00440.0001 5.00460.0001 5.00405.0001 5.00400.0001 5.00450.0001 5.00410.0001 5.00440.0001 5.00460.0001
EM5.00410.0001CS 502922.4 MXN
EM5.00410.0001 CA5.00440.0001 EM5.00400.0001 CA5.00460.0001 CA5.00400.0001 CA5.00405.0001 CA5.00410.0001 CA5.00450.0001 CA5.00420.0001 EM5.00440.0001 EM5.00430.0001 EM5.00405.0001 EM5.00450.0001 CA5.00430.0001 EM5.00420.0001 EM5.00460.0001
Brix Sugar Standards for refractometers, Certipur, MilliporeSigma
Normativas Estándar para índice de refracción
Refractive index standards traceable to SRM from NIST and PTB.
  • Each ampoule contains about 8 ml solution
  • Minimum shelf life of Brix standard 0° is 36 months
  • All other standards minimum shelf life is 24 month
  • Tested for homogeneity and stability
  • Storage at room temperature