Whatman™ Silicone Capmat, Whatman products (Cytiva)
Proveedor: Whatman products (Cytiva)
Whatman™
7704-0105
CA14217-204
Whatman™ Silicone Capmat, Whatman products (Cytiva)
Juntas Sellos de microplaca
Flexible capmats individually seal the top of each well. Capmats may be used on either filter or collection microplates.
  • Pierceable capmats, suitable for autosamplers
  • Resistant to ethanol, methanol, DMSO, and DMF
  • Reseal after needle puncture
Stable at a wide temperature range, capmats can be autoclaved at 120°C or stored at –70°C.