Straight Bore Stopcocks with PTFE Plugs, Chemglass
Proveedor: Chemglass
CG-424-01 CG-424-77 CG-424-71 CG-424-05 CG-424-76 CG-424-03 CG-424-79 CG-424-08 CG-424-04 CG-424-72 CG-424-73 CG-424-75 CG-424-07 CG-424-09
80074-168EA 414.41 MXN
80074-154 80074-134 80074-156 80074-168 80074-146 80074-148 80074-138 80074-160 80074-150 80074-162 80074-140 80074-152 80074-164 80074-142
Straight Bore Stopcocks with PTFE Plugs, Chemglass
Válvulas de cierre
These straight bore stopcocks have a 1:5 taper and solid PTFE plugs.