Hydrion® Jumbo pH Paper, Micro Essential Laboratory
Proveedor: Micro Essential Laboratory
Hydrion®
HJ626 HJ613 HJ610 JR-113 HJ600 HJ633
60786-043CS 4197.73 MXN
60786-043 60786-120 60786-044 60786-040 60786-028 60786-108
Hydrion® Jumbo pH Paper, Micro Essential Laboratory
Papeles de pruebas Tiras indicadoras de pH
Hydrion Jumbo Size pH papers are easier to read with larger color charts
The 0.5" width of the paper enables clearer and quicker color matching. Hydrion Jumbo Insta-Chek Wide Range pH Paper consists of a .5"" by 600"" roll of pH paper, packed in a plastic dispenser with an extra large color chart for easy color matching.