Spectra/Por® 4, Dialysis Membranes, MWCO 12000 to 14000, Spectrum® Laboratories
Proveedor: Spectrum
Spectra/Por®
132700 S632706 132706 132703 132697 132709
25224-375CS 142478.95 MXN
25219-230 25224-375 25219-220 25219-223 28170-160 74400-004
Spectra/Por® 4, Dialysis Membranes, MWCO 12000 to 14000, Spectrum® Laboratories
Tuberías Tuberías para diálisis
Multipurpose dialysis tubing is ideal for general dialysis for a variety of applications and volumes.
  • Hydrophilic, symmetric porosity
Tubing is packaged dry for handling and storage convenience. The tubing is available in 15 m/roll and 30 m/roll to provide better economy for applications requiring larger amounts of tubing.
Dry without glycerin and no soaking is required.