VWR® Ultra Low Temperature Upright Freezers and Freezer Packages, –86° to –50°C
Proveedor: VWR
VWR®
VWR60086DRAKCR EWULTVWR2-3 VWR60086DRAKCO VWR40086DRAKCO VWR32086ARAKCO VWR32086DRAKCO VWR24086V VWR32086DRAKCR EWULTVWR2-1 VWR24086DRAKCR VWR32086ARAKCR VWR40086ARAKCR VWR40086DRAKCR VWR24086ARAKCR VWR40086ARAKCO
10196-234 10002-786 CA10160-908 CA10160-906 10196-232 CA10161-006 CA10161-008 CA10160-900 CA10160-912 CA10160-898 CA10160-910 CA10160-904 CA10160-916 CA10160-902 CA10160-914
VWR® Ultra Low Temperature Upright Freezers and Freezer Packages, –86° to –50°C
Congeladores Ultracongeladores
VWR® Collection Ultra-Low Temperature Upright Freezers are designed and constructed for long-term performance, and they represent the leading choice of laboratory professionals for applications where temperature control is critical.
  • Microprocessor temperature control system
  • Advanced monitor/alarm system
  • Automatic voltage compensator
  • Cascade refrigeration system
  • Non-CFC, non-HFCF, nonflammable refrigerants
  • VWR® Two-year limited parts and labor warranty
  • VWR® Five-year warranty on compressor parts
  • VWR® Five-year limited parts and labor warranty (Canada only)
The VWR® Collection line of ultra-low temperature (ULT) freezers offers a microprocessor temperature control system that maintains constant freezer temperature, along with a sophisticated microprocessor monitor/alarm system.
Modern design includes an easy-to-grasp handle for one-handed operation. Dual wheel, locking swivel casters allow mobility as well as secure positioning. Hinged grille swings out, exposing the foam air filter for easy cleaning or replacement. Industrial-grade cabinets are constructed of high-quality, heavy-gauge, cold-rolled steel. The powder-coated finish is rust- and chip-resistant. RS-232, 4-20 mA analog outputs and one 2.5 cm (1) access port are standard.
Upright freezers feature a triple-point silicone door gasket, three stainless steel shelves that are fully adjustable (bottom of unit acts as fourth shelf), lockable casters, and a vacuum relief port for easy re-entry following door closings. Acoustical mufflers and sound absorbing foam in the freezer base reduce the noise level in the lab. To reduce cold air loss, units feature four inner compartment doors.
The microprocessor temperature control system can be set in 1° increments. The microprocessor monitor/alarm system features audible and visual indicators to keep the operator informed of over/under-temperature conditions, power failures, low battery, and door ajar scenarios. LED indicates actual chamber temperature. Air-cooled, hermetically-sealed dual compressors work in unison and cycle on demand of the microprocessor temperature controller. The automatic voltage compensator detects line frequency, monitors incoming voltage, and adjusts compressor voltage to reduce wear and tear on the compressors.
Certificaciones: Freezers are UL listed, C-UL listed, and CE marked.
Información para pedidos: Exterior clearance requirements: Add 9 cm (3.5") to width for latch/hinge and 18 cm (7") for control panel bezel/wall spacer. Operating range of 115 V units is 103 to 127 V, 120V units is 108 to 130 V, and 230 V units is 208 to 240 V. VWR® Collection –40 °C Upright Freezers and VWR® Collection –80 °C Chest Model Freezers are also available. Contact your Avantor sales representative for additional information.