VWR® Ultra-Low Temperature Chest Freezers, –86 to –50 °C
Proveedor: VWR
VWR®
VWRC2086A VWRC2086D VWRC1386A VWRC0386A VWRC1386D
10160-740EA 483891.42 MXN
10160-740 10160-744 10160-742 10160-738 10160-746
VWR® Ultra-Low Temperature Chest Freezers, –86 to –50 °C
Congeladores Ultracongeladores
Designed and constructed for long-term performance, these freezers represent the leading choice of laboratory professionals for applications where temperature control is critical.
  • Microprocessor temperature control system
  • Advanced monitor/alarm system
  • Automatic voltage compensator
  • Cascade refrigeration system
  • Non-CFC, non-HFCF, nonflammable refrigerants
  • VWR® Two-year limited parts and labor warranty
  • VWR® Five-year warranty on compressor parts
The VWR® Collection line of ultra-low temperature (ULT) freezers offers a microprocessor temperature control system that maintains constant freezer temperature and a sophisticated microprocessor monitor/alarm system.
Modern design includes an easy-to-grasp handle for one-handed operation. Dual wheel, locking swivel casters allow mobility as well as secure positioning. Hinged grille swings out, exposing the foam air filter for easy cleaning or replacement.
The microprocessor temperature control system can be set in 1° increments. LED indicates actual chamber temperature. Air-cooled, hermetically-sealed dual compressors work in unison and cycle on demand of the microprocessor temperature controller. The automatic voltage compensator detects line frequency, monitors incoming voltage, and adjusts compressor voltage to reduce wear and tear on the compressors.
The microprocessor monitor/alarm system features audible and visual indicators to keep the operator informed of over/under-temperature conditions, power failures, low battery, and door ajar.
Chest freezers feature a sleek, low-profile cabinet design that provides easy access to inventory systems. Spring-loaded hinges counterbalance the lid and ensure proper alignment, smooth closure, and a tight seal against the flexible gasket. Polystyrene sub-lids reduce cold air loss.
Certificaciones: Freezers are UL listed, C-UL listed, and CE marked.
Información para pedidos: VWR® Collection Upright –80 °C freezers, see 10160-724 series. For VWR® Collection Upright packages (Racks, back up systems, chart recorder), see 10160-756 series.