WHEATON® Arsine Generator, Modified with One-Piece Scrubber/ Absorber Unit, DWK Life Sciences
Proveedor: DWK Life Sciences
180023 635926 180024
10869-009CS 3359.98 MXN
26316-560 10869-009 10869-012
WHEATON® Arsine Generator, Modified with One-Piece Scrubber/ Absorber Unit, DWK Life Sciences
Digestores Aparato de detección de trazas de elementos
The modified arsine generator with one-piece scrubber or absorber unit eliminates the cumbersome ball and socket joint connecting the scrubber and absorber tubes which conforms to ASTM / USP and EPA specifications
The freestanding system has a standard taper 24/40 Clear-Seal™ joint. It is manufactured from Wheaton 33 low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.