Sampler, MiniSampler, Bürkle
Proveedor: Bürkle
6205-0090 5305-0010 5305-0011 5305-1020 5305-0110 6205-0180 5305-0209 5305-0100
10217-036EA 9708.65 MXN
76047-098 10217-038 10217-032 10217-040 10217-036 10217-044 76047-100 10217-034
Sampler, MiniSampler, Bürkle
Muestreadores Muestreadores de líquidos
The single-use, flexible, suction hose can be quickly replaced, thus ensuring that the samples are never contaminated
If necessary, a new hose can be used for every single sample. Due to its small diameter (8mm) and flexibility, the hose can reach sampling areas which are otherwise inaccessible. Maximum suction lift is five meters, depending on viscosity.
Información accesorios: Accessory sample containers include a screw lid, and are ideal for sampling and transport. Made of PFA for super pure collection of samples, containers are autoclavable.
Información de suministro: Supplied complete with one vacuum pump, 10 PE bottles, PE suction hose, one hose cutter, one hose weight made of stainless steel, and 20 seal-it bottle seals.