VWR® 12" Sieve Pans and Covers
VWR 12" diameter (304.8 mm) pans and covers fit all VWR 12" diameter sieves. Pans are half height (1-5/8") and are available in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available as accessories.