Innova® Open Air Platform Shakers, Eppendorf
Proveedor: Eppendorf
Innova®
M1190-0010 M1191-0030 M1190-0010 M1194-0000 M1194-0010 M1194-0000 M1194-0010 M1191-0000 M1191-0000 M1191-0010 M1191-0020 M1191-0010 M1191-0020
89171-778EA 290250.16 MXN
89171-772 89171-790 CA89171-802 CA89171-778 CA89171-766 89171-802 89171-766 89171-778 CA89171-796 CA89171-784 CA89171-772 89171-784 89171-796
Innova® Open Air Platform Shakers, Eppendorf
Agitadores de laboratorio Agitadores Basculantes y de Rotación
For shaking in ambient conditions: on the bench, in an incubator or in a warm or cold room, the Innova Platform Shakers are the instrument of choice
Quiet operation provides a favorable work enviornment. Shaking speeds between 25  –  500 rpm (±1 rpm) provides versatility for culturing a wide range of cell types. Includes audible and visual alarms, timer (0  –  99.9 h) with continuous operation or automatic shut-off at the end of the study.
Dependable operation is due in large part to the New Brunswick triple-eccentric counterbalanced drive. The Innova platform shaker model 2050 comes with additional stabilizing feet and counterweight to balance larger loads on an oversized platform. Triple-eccentric counterbalanced drive in cast iron housing provides vibration and trouble free operation for years.
Información para pedidos: Platforms for all models are sold separately. Versatile accessory platform (sold separately) accommodates larger flasks.