Septa GP, SGE
Proveedor: Trajan Scientific and Medical
041821 0418240 041825 041824 041823 041826 041828 041820 041822 041829 041827
60361-502PK 1386.37 MXN
60361-506 60361-504 60361-494 60361-492 60361-508 60361-510 60361-498 60361-496 60361-502 60361-512 60361-500
Septa GP, SGE
Septos
Septa are designed to reliably seal against the carrier gas pressure in the inlet.
  • Good durability
  • Resealing, and tear resistance properties
  • Excellent solvent resistance
Septa GP combines significantly longer injection life, low bleed, and low injection port adhesion. Intended for low temperature applications, they are blister packed to prevent absorption from the environment.
All septa should be capable of being pierced and resealed time after time, without pieces of the septa being deposited in the GC inlet system. It is essential that the septa does not contaminate or bleed material into the chromatographic system.
Certificaciones: RoHS compliant per Directive 2011/65/EU.