Septa HT, SGE
Proveedor: Trajan Scientific and Medical
041898 041895 041897 0418990 041899 0418992 041896 041881 041883
89205-430PK 1833.59 MXN
89205-430 60361-526 89205-434 89205-432 89205-428 89205-438 89205-426 89205-436 60361-522
Septa HT, SGE
Septos
Septa are designed to reliably seal against the carrier gas pressure in the inlet.
  • Excellent durability
  • Resealing
  • Tear resistance
  • Solvent resistance
Septa HT are bleed and temperature optimized, and combined with outstanding mechanical properties, they are ideally suited for the highest temperature applications. They retain softness and pierceability at high temperatures, in addition to low injection port adhesion. Septa HT are blister packed to prevent absorption from the environment.
All septa operate to a maximum temperature of 400 °C (752 °F), with the exception of the 17 mm sizes, which offer a maximum temperature of 330 °C (626 °F).
All septa should be capable of being pierced and resealed time after time, without pieces of the septa being deposited in the GC inlet system. It is essential that the septa does not contaminate or bleed material into the chromatographic system.
Certificaciones: RoHS compliant per Directive 2011/65/EU.