Septa EC, SGE
Proveedor: Trajan Scientific and Medical
041904 041906 041902 041905 041900 041912 041843 041901 041845 041841 041903 041842
89205-416PK 1654.7 MXN
89205-420 14225-836 14225-826 89205-424 89205-412 89205-422 10799-700 89205-416 14225-830 89205-414 89205-418 14225-824
Septa EC, SGE
Septos
Septa are designed to reliably seal against the carrier gas pressure in the inlet.
  • Excellent durability
  • Resealing
  • Tear resistance
  • Solvent resistance
Septa EC combines significantly longer injection life, low bleed, and low injection port adhesion. Intended for high turnover applications, they are blister packed to prevent absorption from the environment.
All septa operate to a maximum temperature of 400 °C (752 °F), with the exception of the 17 mm sizes, which offer a maximum temperature of 330 °C (626 °F).
All septa should be capable of being pierced and resealed time after time, without pieces of the septa being deposited in the GC inlet system. It is essential that the septa does not contaminate or bleed material into the chromatographic system.
Certificaciones: RoHS compliant per Directive 2011/65/EU.