PakSense™ XpressPDF™ Temperature Dataloggers, Thermco®
Proveedor: Thermco
XPDF5528 XPDF551525 XPDF5528-1 XPDF551525-1
89412-414CS 10764 MXN
89412-408 89412-410 89412-414 89412-412
PakSense™ XpressPDF™ Temperature Dataloggers, Thermco®
Registradores de datos
Designed to allow simplistic operation, PakSense™ XpressPDF™ Labels eliminate the need for reading devices and proprietary download software
These temperature loggers are optimal for applications that require fully independent operation, such as global shipments. Labels feature an integrated USB connection point which can be plugged directly into the USB port of a computer for data generation. Three options are available, including a highly accurate model for use in the life sciences industry, a general-use model, and a basic version.
Certificaciones: UL listed. CE marked.
Información para pedidos: Accessories not included.