Heated Incubators, 7002 Series, Caron Products
Proveedor: Caron Products
7002-10-2 7002-25-2 7002-33-2 7002-10-3 7002-25-1 7002-10-1 7002-33-1 7002-33-3 7002-25-3
10216-776EA 391779.15 MXN
10216-768 10216-764 10216-776 10216-766 10216-778 10216-772 10216-774 10216-780 10216-770
Heated Incubators, 7002 Series, Caron Products
Incubadoras Incubadores estándar
Caron’s Heated Incubators offer controlled temperature conditions (10°C above ambient to 70°C)
115V units will plug into a standard laboratory electrical outlet, contributing to cost savings and energy efficiency. Temperature uniformity is excellent for reliable, repeatable results. Key standard features include: touchscreen user interface; built-in preventative maintenance; tool-less removable interior; sliding shelves; horizontal airflow system that generates gentle and evenly distributed airflow across all shelf locations; heated, triple pane glass door (708L and 934 L models); and Caron green door handles are aesthetically pleasing and enhance ergonomics.
Powder-coated cold rolled steel exterior construction.
Note: View the Product Accessories List, in Datasheets under Related Information. To order specific accessories, search by part number.
These incubators offer consistently reliable results, best-in-class customer-friendly features and energy efficiency.
Información para pedidos: Accessories and Options not included. Popular options include: remote alarm contacts (ALRM302) and the built-in temperature recorder (RCDR316). 1-year parts and labor warranty standard. 283 L units (10 ft3) come standard with leveling feet and three (3) shelves. 708 L (25 ft3) units come standard with casters/leveling feet combo and four (4) shelves. 934 L (33 ft3) units come standard with casters/leveling feet combo and five (5) shelves.