Unions for Gas Chromatography, Trajan Scientific and Medical
Proveedor: Trajan Scientific and Medical
103402 1236300 103406 103403 101300 1034030 1034200 103420 1034183 101290 103404 103407 101230 101270 101340 103418 18500002 103408 1236306 103400 1034605 1034050 103405 1236302 103461 1034604 1034180 1034187 103330
14226-528PK 1128 MXN
10799-802 10799-804 10799-800 10799-806 14226-508 10799-808 14226-506 14226-528 14226-514 14226-534 14226-532 14226-530 14226-570 14227-500 14227-502 10799-798 14226-518 14226-538 14226-526 14226-504 14226-524 14226-568 14226-546 14226-522 14226-544 14226-500 14226-566 14226-520 14226-540
Unions for Gas Chromatography, Trajan Scientific and Medical
Racores y conectores
Stainless steel unions, tees and crosses come in a variety of tube sizes and configurations, such as straight through, reducing, tees, crosses and bulkheads
GLT unions are inert and mainly used for connecting fused silica tubing of various internal diameters. All SGE unions are supplied with the appropriate nuts, ferrules and/or sealing rings. Replacement nuts are also available.
Certificaciones: RoHS compliant per Directive 2011/65/EU
Información para pedidos: Unions and nuts are available in a range of sizes to connect 1/4, 1/8, and 1/16" tubing and fused silica.