Recirculating Water Pump, JP, VELP Scientifica
Proveedor: VELP Scientifica
F307A0660 F30630198
10184-076EA 105480.65 MXN
10184-076 10184-074
Recirculating Water Pump, JP, VELP Scientifica
Bombas Bombas para bidón
Recirculating Water Pump that ensures optimum aspiration depending on the digestion phases and the number of samples placed in the digester connected to the pump.
  • Optimized Performance
  • Robust Design
  • Practical
Recirculating Water Pump that ensures optimum aspiration depending on the digestion phases and the number of samples placed in the digester connected to the pump.
Optimized Performance–Aspiration matches digestion phases and sample number. Eliminates the problem of unreliable water supply. Up to 35 liters/minute airflow pumping speed. Selectable performance mode related to samples.
Información para pedidos: Complete your preferred system with the SMS Scrubber that neutralizes all the fumes. Two JP Pumps are recommended when using DK and DKL 42/26.