Scrubber, SMS, VELP Scientifica
Proveedor: VELP Scientifica
F307A0660
10184-078EA 56585.59 MXN
10184-078
Scrubber, SMS, VELP Scientifica
Digestores Estaciones de trabajo de mineralización
Designed to neutralize corrosive and toxic fumes produced during oxidative mineralizations or similar processes.
  • Complete system
  • Convenient to use
Neutralization with acids or bases. Optional absorption with activated carbon. Ideal for high fume levels such as with soybean analysis. Condensate flasks removable for ease of emptying. Easy to replace the neutralizing reagents. Small footprint.
Información para pedidos: Complete your preferred system with JP Recirculating Water Pump that aspirates fumes.