Gel Refrigerant Packs, Therapak®
Proveedor: Therapak
56400 56407
10029-374CS 3193.02 MXN
10029-374 10029-372
Gel Refrigerant Packs, Therapak®
Envases fríos
Gel Refrigerant Packs, 0°C (32°F), are an economical alternative to wet ice commonly used as cold packs in conjunction with insulated shippers for transporting laboratory specimens.
  • Reliable Refrigeration
  • Safe Transport
  • No Leaks or Spills
  • Durable 4⅟₁₀ mil Polyethylene Film Encloses Packs
  • Economical Alternative to Wet Ice
Gel refrigerant packs provide reliable refrigeration for safe transport without leaks or spills.
A durable 4⅟₁₀ mil polyethylene film encloses the packs.