Multifuge™ X1/X1R Benchtop Centrifuges, Thermo Scientific
Proveedor: Thermo Scientific
75004211 75112024 75518120 75118120 75512419 75512024 75004251 75118119 75112419 75518382 75518119 75118382 75512963
97039-592EA 634489.6 MXN
97039-578 10304-274 97039-592 10304-272 10304-278 97039-266 10304-268 10304-276 10304-266 97039-576 10304-270 97039-270 10304-280
Multifuge™ X1/X1R Benchtop Centrifuges, Thermo Scientific
Centrífugas Centrífugas de sobremesa
The Multifuge™ X1 Centrifuge series offers industry-leading throughput, the most versatile range of rotors and adapters, and energy-efficient operation. Recommended for use in cell culture, microplate, bioproduction, food safety, blood separation, bodily fluid processing, pharmaceutical, biotechnology, stem cell, microbiology, molecular biology, proteomics,and genomics applications. This centrifuge series offers unparalleled processing performance, maximum versatility, and exceptional energy efficiency. A broad range of rotors and adapters, including lightweight Fiberlite carbon fiber rotors, are available to handle such diverse laboratory applications.
To facilitate simple operation, each unit features an intuitive control panel and digital display
The Auto-Lock™ III Rotor System is a secure locking system that allows easy push-button installation and exchange of rotors. The system saves time in between applications, and offers the user easy access to the chamber, enabling quick cleaning, a healthier working environment, and longer unit life.
The SMARTSpin™ Technology is an advanced rotor management system which maximizes acceleration, braking and residual load imbalance for each rotor and bucket option. By this technique, system optimizes safety and improves separations.
Detailed operation information, including display of set and actual parameters, maximize run performance and temperature accuracy for improved sample protection. The user can easily adjust speed, time, and temperature, even during the run, with a convenient keyboard for direct input. Innovative lid locking technology enables one finger downward motion for simple closing and locking of the centrifuge lid, even on tall benches (with optional automatic lid opening). A double temperature management system displays the air temperature in the chamber as well as the sample temperature, ensuring accurate control of the sample temperature during the run.
Certificaciones: UL listed. CE marked. CSA certified. Certified Biosafety.
Información para pedidos: Accessories are not included.