VWR® Sterile Coveralls, 10⁻⁶ SAL
Proveedor: VWR
VWR®
89523-472 89523-460 89523-466 89523-462 89523-468 10853-872 89523-458 89523-470 89523-464
89523-464CS 3299.74 MXN
89523-464 89523-462 89523-458 89523-468 89523-466 89523-472 89523-460 10853-872 89523-470
VWR® Sterile Coveralls, 10⁻⁶ SAL
Prendas desechables
VWR® new sterile coveralls provide a solid option for customers requiring a functional garment that is 10-6 SAL
  • Gamma irradiated to SAL 10-6
  • Durable fabric
  • Excellent breathability
  • Suitable for ISO Class 5 or higher cleanrooms
  • Individually bagged
These comfortable coveralls are manufactured from a durable SMS fabric and feature bound seams. Rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Coveralls are Clean Level II certified according IEST test results, and offer a BFE of over 98%. Designed for performance in a variety of clean environments in multiple markets including Compounding Pharmacies and Biopharmaceutical production.
Certificaciones: Manufactured in an ISO Class 8 (FED-STD-209E Class 100,000/M3.5) cleanroom.