Extran® AP 41 Powdered Enzymatic Cleaner for Automatic Washing, MilliporeSigma
1.07570.9025 1.07570.9025
EM1.07570.9025 CA1.07570.9025
Extran® AP 41 Powdered Enzymatic Cleaner for Automatic Washing, MilliporeSigma
When diluted with water, concentrates can clean by themselves without leaving residues
  • 1.07570.2000 is offered as an alternative to EX0992-1. Different Pack Size, Same Specifications.
  • 1.07570.9025 replaces and is the same as EX0992-7. Part # Change only, Same Pack Size, Same Specifications.
  • 1.07570.9025 is offered as an alternative to EX0992-5. Different Pack Size, Same Specifications.
For routine laboratory applications, no scrubbing is necessary once items are removed from the cleaning bath. For best results, items to be cleaned should be soaked for 2 to 24 hours, and rinsed with distilled water immediately after removal from cleaning bath.