MColortest™ Ammonium Test, MilliporeSigma
Proveedor: MilliporeSigma
MColortest™
1.08024.0001 1.08024.0001
EM1.08024.0001EA 4470.44 MXN
CA1.08024.0001 EM1.08024.0001
MColortest™ Ammonium Test, MilliporeSigma
Kits para test medioambiente
The test measures both ammonium ions and dissolved ammonia in e.g
groundwater and surface water, seawater, drinking water and aquarium water. The test kit contains the easy to use colour card and the sliding comparator.