Aura™ 9210+ N95 Particulate Respirators, 3M™
Proveedor: 3M
7100110559
10031-464CS 10017.24 MXN
10031-464
Aura™ 9210+ N95 Particulate Respirators, 3M™
Respiradores Mascarillas desechables Mascarillas para partículas
This light weight three-panel, flat-fold disposable respirator with its innovative design helps provide comfortable, reliable worker protection against non-oil based particles.
  • Lightweight, 3-panel, flat fold, disposable
  • Protection against non-oil based particles
  • Ideal for grinding, sanding, sweeping, bagging and other dusty or arid operations
Its comfortable braided headbands help to keep respirator securely in place and minimize pulling of hair
Has wide range of applications like Grinding, Sanding, Sweeping, Bagging and other dusty or arid operations.
Precaución: These respirators help reduce exposures to certain airborne particulates. Before use, the wearer must read and understand the User Instructions provided as a part of the product packaging. A written respiratory protection program must be implemented meeting all the requirements of OSHA 1910.134 including training, fit testing and medical evaluation.