Compact Digital Microplate Shaker, Thermo Scientific
Proveedor: Thermo Scientific
88880023
10054-208EA 27158.63 MXN
10054-208
Compact Digital Microplate Shaker, Thermo Scientific
Agitadores de laboratorio Agitadores de Microplaca
Precise and accurate microplate shakers capable of shaking up to 4 microplates in one run
  • PID control ensures consistent shaking results
  • Digital display for easy viewing
  • Compact design is ideal for placing in incubators (non-CO2), chambers and refrigerators (up to 60 °C)
  • Automatic Overload Shutdown > 4.4 kg (9.7 lbs)
Ideal for immunoassay, screening and magnetic bead applications.
Certificaciones: CE marked.