Comply™ Thermalog Steam Chemical Integrator, 3M™
Proveedor: 3M
MMM2134MM
34010-086CS 13772.22 MXN
34010-086
Comply™ Thermalog Steam Chemical Integrator, 3M™
Indicadores de esterilización
Class 5 steam chemical integrators can be used in all 250 to 275 °F steam sterilization cycles.
  • Immediate and accurate method of monitoring the sterilization
  • Ink technology provides easy to read safe or unsafe result
Certificaciones: ANSI/AAMI/ISO 11140-1:2005 Class 5.