Bottle Brush, Micronova
Proveedor: Micronova
BRWH-67
89187-752CS 18483.94 MXN
89187-752
Bottle Brush, Micronova
This wirewound brush features a smaller diameter handle and softer bristles for delicate cleaning of jars and bottles with smaller openings.
Base resins used to produce block and bristles are made from FDA approved materials. BPA free.