20 mm Headspace Vials, Thermo Scientific
Proveedor: Thermo Scientific
C4020-143 C4020-2 C4020-25 C4020-6 C4020-60 C4020-410 C4020-210 C4020-27 C4020-S10 C4020-S20 C4020-20 C4020-10
66064-346CS 13262.08 MXN
66065-358 66064-346 66065-348 66030-782 66065-328 66065-344 66064-350 66065-340 69400-116 69400-118 66064-348 66030-784
20 mm Headspace Vials, Thermo Scientific
Viales Viales para cromatografía
Thermo Scientific National Headspace Vials meet or exceed OEM specifications.
Vials feature a beveled crimp finish which gives a much tighter seal than flat crimp surfaces. Manufactured from clear 33 expansion borosilicate glass. Flat-bottom vials feature a slightly longer neck, providing a custom fit for Carlo Erba and Agilent® autosamplers. Round-bottom vials are designed to fit PerkinElmer®, Tekmar®, and Varian® headspace autosamplers. Seals have attached septa, unless otherwise specified.
Thermo Scientific National Unassembled Headspace Convenience Kits are available for the most popular vial and seal combinations. Includes 100 vials and 100 seals with prefitted septa. 20 ml round bottom vials.
Clear vials are made of Type 1 borosilicate glass with 20 mm crimp seal finish and are designed to fit most headspace autosamplers. Flat bottom vials maximize heating efficiency when used with bottom. Round bottom vials are more easily handled by robotic arms that lift the vial from the tray. Round bottom vials distribute the internal pressure created at high temperatures across the glass surface. Beveled edge finish features a sturdy rim that presses into the septum for a more effective seal.