VWR® Disposable Hexagonal Weigh Boats
Proveedor: VWR
VWR®
HS14251CV HS14251BV HS14251AV HS14251DV
10770-456CS 9123.21 MXN
10770-456 10770-458 10770-452 10770-454
VWR® Disposable Hexagonal Weigh Boats
Navecillas Navecillas de pesaje
These polystyrene weighing boats feature smooth surfaces to enable accurate pour-out with minimal sample loss.
  • Flat bottom avoids tipping
  • Sloping sides provide accurate transfer
  • Antistatic
  • Temperature resistant up to 70°C (158°F)
Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols and bases. These flexible, antistatic boats bend at points to form pouring spouts.